Learn More
Thermo Scientific Chemicals L-Aspartyl-L-phenylalanine methyl ester, 98%
Catalog Number p-3735544
Click to view available options
Quantità:
1 g
5 g
25 g
Imballaggio:
Glass Bottle
22839-47-0
C14H18N2O5
294.31
MFCD00002724
IAOZJIPTCAWIRG-QWRGUYRKSA-N
aspartame, nutrasweet, asp-phe-ome, asp-phe methyl ester, aspartam, aspartamum, aspartamo, l-aspartyl-l-phenylalanine methyl ester, aspartylphenylalanine methyl ester, dipeptide sweetener
134601
CHEBI:2877
(3S)-3-amino-4-[[(2S)-1-methoxy-1-oxo-3-phenylpropan-2-yl]amino]-4-oxobutanoic acid
COC(=O)[C@H](CC1=CC=CC=C1)NC(=O)[C@@H](N)CC(O)=O
22839-47-0
C14H18N2O5
294.31
MFCD00002724
IAOZJIPTCAWIRG-QWRGUYRKSA-N
aspartame, nutrasweet, asp-phe-ome, asp-phe methyl ester, aspartam, aspartamum, aspartamo, l-aspartyl-l-phenylalanine methyl ester, aspartylphenylalanine methyl ester, dipeptide sweetener
134601
CHEBI:2877
(3S)-3-amino-4-[[(2S)-1-methoxy-1-oxo-3-phenylpropan-2-yl]amino]-4-oxobutanoic acid
COC(=O)[C@H](CC1=CC=CC=C1)NC(=O)[C@@H](N)CC(O)=O
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Materiale o nome chimico L-Aspartyl-L-phenylalanine methyl ester
Punto di fusione 248.0°C to 250.0°C
Colore White
Intervallo percentuale saggio 98%
Spettro a infrarossi Authentic
Formula lineare HO2CCH2CH(NH2)CONHCH(CH2C6H5)CO2CH3
Perdita all'essiccatura 4.5% max. (105°C, 4 hrs)
Indice di Merck 15, 829
Imballaggio Glass Bottle
Informazioni di solubilità Solubility in water: sparingly soluble.
Rotazione specifica + 12.50
Peso formulazione 294.3
Trasmittanza 95% min. (c=1, 2N HCl, 430nm) 1cm cell
Percent Purity 98%
Quantità 1 g
Forma fisica Crystalline Powder