Learn More
Masterflex™ L/S™ Standard Pump Head Replacement End Bell Assembly
Masterflex™ L/S™ Standard Pump Head Replacement End Bell Assemblies extend the operating life of standard pump heads
Marca: Masterflex™ 07015-81
Dettagli aggiuntivi : Peso : 0.80000kg
Descrizione
- Replacement parts to keep pump head operating for an extended period of time
- Two end bell assemblies are required for a complete head assembly
- Choose replacement parts suitable for your I/P™ Standard pump head size
- Suitable for use with L/S™ Standard Pump Heads
Specifica
CRS Rotor, L/S 15 Tubing | |
L/S™ Standard Pump Heads 07015-00, 07015-20, 07015-70, and 07015-72 | |
Replacement End Bell Assembly |
Polycarbonate | |
L/S 15 |