missing translation for 'onlineSavingsMsg'
Learn More

CD117/c-Kit/SCF-Receptor Rabbit Polyclonal Antibody, Epredia™

Optimize staining of formalin-fixed, paraffin-embedded tissues with Epredia™ CD117/c-Kit/SCF-Receptor, Rabbit Polyclonal Antibody.

Marca:  Epredia RB-9038-P0

Dettagli aggiuntivi : Peso : 0.00170kg

 vedi altre versioni di questo prodotto

Codice prodotto. 12663387

  • 218.00€ / 100µL
Spedizione stimata: 21-05-2024
per visualizzare lo stock disponibile

Effettua il per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso!


Descrizione

Descrizione

Recognizes a protein of 145kDa, which is identified as CD117/p145kit. This rabbit polyclonal antibody does not interfere with the binding of SCF to c-kit. It precipitates both the unoccupied as well as the occupied form of c-kit. The binding of the stem cell factor (SCF) to the c-kit-encoded receptor tyrosine kinase (Type III) stimulates a variety of biochemical responses that culminate in cellular proliferation, migration, or survival. C-kit plays an important role in hematopoiesis, melanogenesis, and gametogenesis. This antibody is excellent for staining of formalin-fixed, paraffin-embedded tissues.

Host Species: Rabbit

Isotype: IgG

Species Reactivity: Human and Dog. Others not tested

Epitope: Cytoplasmic domain

Immunogen: A synthetic peptide from the cytoplasmic domain of human CD117/c-kit protein

Molecular Weight: 145 kDa

Positive Control: Stromal tumor, skin or tonsil

Cellular Localization: Membrane

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
Specifica

Specifica

CD117/c-Kit/SCF-Receptor
Polyclonal
200µg/mL
Rabbit
100 μL
Primary
Concentrated
Immunohistochemistry (Paraffin)
Polyclonal
Unconjugated
A synthetic peptide from the cytoplasmic domain of human CD117 / c-kit protein
IVD
Canine
IgG
Suggerimenti di prodotto

Suggerimenti di prodotto

Videos
SDS
Documenti

Documenti

Certificati
Offerte speciali

Offerte speciali