missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ Lab Vision™ CD3ε Rabbit Monoclonal Antibody

Obtain accurate and reproducible results in immunohistochemistry applications with Epredia™ CD3ε, Rabbit Monoclonal Antibody.

Marca:  Epredia™ RM-2109-S

Dettagli aggiuntivi : Peso : 0.00250kg

 vedi altre versioni di questo prodotto

Codice prodotto. 12633637

  • 1860.00€ / 1mL
Spedizione stimata: 16-05-2024
per visualizzare lo stock disponibile

Effettua il per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso!


Descrizione

Descrizione

CD3 consists of five different polypeptide chains (designated as gamma, delta, epsilon, zeta, and eta). The CD3 complex is closely associated at the lymphocyte cell surface with the T cell antigen receptor (TCR). It is involved in signal transduction to the T cell interior following antigen recognition. CD3 antigen is a highly specific marker for T cells, and is present in the majority of T cell neoplasms.

Host Species: Rabbit

Clone: EP449E

Isotype: IgG

Species Reactivity: Human. Others not known.

Immunogen: A synthetic peptide corresponding to residues in the cytoplasmic domain of human CD3ε. The antibody does not cross react with other subunits of CD3.

Molecular Weight: 23kDa

Positive Control: Tonsil

Cellular Localization: Membrane

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
Specifica

Specifica

CD3e
Monoclonal
Unconjugated
A synthetic peptide corresponding to residues in the cytoplasmic domain of human CD3epsilon. The antibody does not cross react with other subunits of CD3.
IVD
Primary
Concentrated
Immunohistochemistry (Paraffin)
EP449E
Rabbit
1 mL
Cancer and Tumor Biology
Human
IgG
Suggerimenti di prodotto

Suggerimenti di prodotto

Videos
SDS
Documenti

Documenti

Certificati
Offerte speciali

Offerte speciali