Learn More
Epredia™ Lab Vision™ CEA (Carcinoembryonic Antigen)/CD66e Ab-2 Rabbit Polyclonal Antibody with BSA and azide
Ensure accurate, reproducible results in immunohistochemistry, western blotting and immunoprecipitation experiments.
Marca: Epredia™ RB-368-A0
Dettagli aggiuntivi : Peso : 0.00170kg
Descrizione
Carcinoembryonic antigen (CEA), is synthesized during development in the fetal gut, and is re-expressed in increased amounts in intestinal carcinomas and several other tumors. Antibody to CEA is reportedly useful in identifying the origin of various metastatic adenocarcinomas and in distinguishing pulmonary adenocarcinomas (60-70% are CEA+) from pleural mesotheliomas (rarely or weakly CEA+).Host Species: Rabbit
Isotype: IgG
Species Reactivity: Human and Mouse. Weakly reacts with Rat. Does not react with dog. Others not known
Immunogen: Human CEA isolated from hepatic metastasis of colon adenocarcinoma
Molecular Weight: 180 kDa
Positive Control: LS174T cells or colon CA
Cellular Localization: Cytoplasmic and luminal membrane
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
- Western Blotting
- Immunoprecipitation (Native and denatured)
Specifica
CEA (Carcinoembryonic Antigen)/CD66e Ab-2 | |
Polyclonal | |
Rabbit | |
LS174T cells or colon CA | |
IVD | |
Primary | |
Concentrated |
Immunoprecipitation, Western Blot | |
Unconjugated | |
Human CEA isolated from hepatic metastasis of colon adenocarcinoma | |
100 μL | |
Oncogenesis | |
Human | |
IgG |