missing translation for 'onlineSavingsMsg'
Learn More

Cytokeratin 19 Ab-5 Mouse Monoclonal Antibody, Epredia™

Ensure accurate, reproducible results in immunohistochemistry and western blotting procedures with Epredia™ Cytokeratin 19 Ab-5, Mouse Monoclonal Antibody.

Marca:  Epredia MS-1902-R7

Dettagli aggiuntivi : Peso : 0.01200kg

 vedi altre versioni di questo prodotto

Codice prodotto. 12610447

Questo articolo non è attualmente disponibile o è stato dismesso.
La preghiamo di consultare la pagina del prodotto per individuare le possibili alternative.


Descrizione

Descrizione

Cytokeratin 19 is a member of type I acidic subfamily of intermediate filaments. It is expressed in various human tissues except in liver. Keratin 19 is not expressed in hepatocytes, therefore, antibody to keratin 19 is useful in the identification of liver metastasis.

Host Species: Mouse

Clone: RCK108

Isotype: IgG1

Species Reactivity: Human. Others not known.

Epitope: Not determined

Immunogen: Cytoskeleton preparation of T24 cells

Molecular Weight: 40 kDa

Positive Control: LS174T and HT29 cells, Colon CA

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting

Note:

USA: RUO; Int'l: RUO

Specifica

Specifica

Cytokeratin 19 Ab-5
Monoclonal
Unconjugated
Cytoskeleton preparation of T24 cells
RUO
Primary
Prediluted
Immunohistochemistry (Paraffin), Western Blot
RCK108
Mouse
7 mL
Cytoskeletal Protein
Human
IgG1
Suggerimenti di prodotto

Suggerimenti di prodotto

Videos
SDS
Documenti

Documenti

Certificati
Offerte speciali

Offerte speciali