Learn More
Cytokeratin 19 Ab-5 Mouse Monoclonal Antibody, Epredia™
Ensure accurate, reproducible results in immunohistochemistry and western blotting procedures with Epredia™ Cytokeratin 19 Ab-5, Mouse Monoclonal Antibody.
Marca: Epredia MS-1902-PABX
Dettagli aggiuntivi : Peso : 1.00000kg
Descrizione
Cytokeratin 19 is a member of type I acidic subfamily of intermediate filaments. It is expressed in various human tissues except in liver. Keratin 19 is not expressed in hepatocytes, therefore, antibody to keratin 19 is useful in the identification of liver metastasis.Host Species: Mouse
Clone: RCK108
Isotype: IgG1
Species Reactivity: Human. Others not known.
Epitope: Not determined
Immunogen: Cytoskeleton preparation of T24 cells
Molecular Weight: 40 kDa
Positive Control: LS174T and HT29 cells, Colon CA
Cellular Localization: Cytoplasmic
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
- Western Blotting
Note:
USA: RUO; Int'l: RUO
Specifica
Cytokeratin 19 Ab-5 | |
Monoclonal | |
1mg/mL | |
Mouse | |
200 μL | |
Cytoskeletal Protein | |
Human | |
IgG1 |
Immunohistochemistry (Paraffin), Western Blot | |
RCK108 | |
Unconjugated | |
Cytoskeleton preparation of T24 cells | |
RUO | |
Primary | |
Concentrated |