Learn More
Epredia™ Lab Vision™ Cytokeratin 8 Rabbit Monoclonal Antibody
Obtain accurate, reproducible results in immunohistochemistry experiments with Epredia™ Cytokeratin 8, Rabbit Monoclonal Antibody.
Marca: Epredia™ RM-2107-S0
Descrizione
Cytokeratin 8 belongs to the type B (basic) subfamily of low molecular weight keratins and usually exists in combination with cytokeratin 18. Cytokeratin 8 is primarily found in the non-squamous simple epithelia. It is expressed in mesothelioma and most adenocarcinomas of simple epithelium including breast ducts, gall bladder, intestine, liver, pancreas and prostate.Host Species: Rabbit
Clone: EP1628Y
Isotype: IgG
Species Reactivity: Human. Others not known.
Immunogen: A synthetic peptide corresponding to residues on the C-terminus of human CK8
Molecular Weight: 52 kDa
Positive Control: Pancreas, Mesothelioma
Cellular Localization: Cytoplasmic
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
Specifica
| Cytokeratin 8 | |
| Monoclonal | |
| Unconjugated | |
| A synthetic peptide corresponding to residues of the human CK18 | |
| IVD | |
| Primary | |
| Concentrated |
| Immunohistochemistry (Paraffin) | |
| EP1628Y | |
| Rabbit | |
| 100 μL | |
| Cancer and Tumor Biology | |
| Human | |
| IgG |
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.