Learn More
Epredia™ Lab Vision™ Cytokeratin 8 Rabbit Monoclonal Antibody
Obtain accurate, reproducible results in immunohistochemistry experiments with Epredia™ Cytokeratin 8, Rabbit Monoclonal Antibody.
313.14€ - 2135.00€
Specifica
| Antigene | Cytokeratin 8 |
|---|---|
| Clone | EP1628Y |
| Applicazioni | Immunohistochemistry (Paraffin) |
| Classificazione | Monoclonal |
| Coniugato | Unconjugated |
| Codice del prodotto | Marca | Quantità | Forma | Prezzo | Quantità e disponibilità | ||||
|---|---|---|---|---|---|---|---|---|---|
| Codice del prodotto | Marca | Quantità | Forma | Prezzo | Quantità e disponibilità | ||||
|
12653627
|
Epredia™
RM-2107-S0 |
100 μL | Concentrated | ||||||
|
12633627
|
Epredia™
RM-2107-S |
1 mL | Concentrated | ||||||
|
12613627
|
Epredia™
RM-2107-R7 |
7 mL | Prediluted | ||||||
|
12643627
|
Epredia™
RM-2107-RQ |
12 mL | Ready-to-Use | ||||||
Descrizione
Cytokeratin 8 belongs to the type B (basic) subfamily of low molecular weight keratins and usually exists in combination with cytokeratin 18. Cytokeratin 8 is primarily found in the non-squamous simple epithelia. It is expressed in mesothelioma and most adenocarcinomas of simple epithelium including breast ducts, gall bladder, intestine, liver, pancreas and prostate.Host Species: Rabbit
Clone: EP1628Y
Isotype: IgG
Species Reactivity: Human. Others not known.
Immunogen: A synthetic peptide corresponding to residues on the C-terminus of human CK8
Molecular Weight: 52 kDa
Positive Control: Pancreas, Mesothelioma
Cellular Localization: Cytoplasmic
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
Specifica
| Cytokeratin 8 | |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| IVD | |
| A synthetic peptide corresponding to residues of the human CK18 |
| EP1628Y | |
| Monoclonal | |
| Rabbit | |
| Cancer and Tumor Biology | |
| Primary |
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.