missing translation for 'onlineSavingsMsg'
Learn More

HER-2/c-erbB-2/neu5 Ab-15 Mouse Monoclonal Antibody, Epredia™

Recommended for Immunohistochemistry, Western Blotting and Immunoprecipitation (Native and Denatured), Epredia™ HER-2/c-erbB-2/neu5 Ab-15, Mouse Monoclonal Antibody provides accurate, reproducible results.

Marca:  Epredia MS-599-P1ABX

Dettagli aggiuntivi : Peso : 1.00000kg

 vedi altre versioni di questo prodotto

Codice prodotto. 12651727

  • 670.00€ / 100µL
Spedizione stimata: 21-05-2024
per visualizzare lo stock disponibile

Effettua il per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso!


Descrizione

Descrizione

c-erbB-2 is a receptor tyrosine kinase of the c-erbB family. Approximately 25% of primary breast and ovarian tumors were found to overexpress the protein

Host Species: Mouse

Clone: 3B5

Isotype: IgG1

Species Reactivity: Human, Monkey, Mouse and Rat. Others not tested.

Epitope: C-terminus

Immunogen: A synthetic peptide from the C-terminus of human HER-2 protein. This sequence is identical in rat neu protein.

Molecular Weight: 185 kDa

Positive Control: SKBR-3 or MAD109 cells, or Breast CA

Cellular Localization: Membrane

Recommended for:

  • Immunohistochemistry
  • Western Blotting
  • Immunoprecipitation (Native and Denatured)

Note:

USA: RUO; Int'l: RUO

Specifica

Specifica

HER-2/c-erbB-2/neu5 Ab-15
Monoclonal
1mg/mL
Mouse
100 μL
EGFR Family
Human, Mouse, Non-human Primate, Rat
IgG1
Immunoprecipitation, Western Blot
3B5
Unconjugated
A synthetic peptide from the C-terminus of human c-erbB-2 protein. This Sequence Is Identical In Rat Neu Protein.
RUO
Primary
Concentrated
Suggerimenti di prodotto

Suggerimenti di prodotto

Videos
SDS
Documenti

Documenti

Certificati
Offerte speciali

Offerte speciali