missing translation for 'onlineSavingsMsg'
Learn More

Mucin 3 (MUC3) Ab-2 Mouse Monoclonal Antibody, Epredia™

Epredia™ Mucin 3 (MUC3) Ab-2, Mouse Monoclonal Antibody provides accurate, reproducible results.

Marca:  Epredia MS-1734-P0

 vedi altre versioni di questo prodotto

Codice prodotto. 12600097

Questo articolo non è attualmente disponibile o è stato dismesso.
La preghiamo di consultare la pagina del prodotto per individuare le possibili alternative.
Visualizzare prodotti alternativi »

Questo articolo non è restituibile. Consulta la politica di reso

Descrizione

Descrizione

Mucin 3 (MUC3) is a glycoprotein. Distributed in colon and rectum, and to a lesser extent in breast, lung and salivary gland tissues.

Host Species: Mouse

Clone: 1143/B7

Isotype: IgG2a

Species Reactivity: Human. Others not known.

Epitope: Tandem repeats of the human MUC3

Immunogen: MUC3 tandem repeat peptide

Molecular Weight: High

Positive Control: Colon CA

Cellular Localization: Membrane

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)

Note:

USA: RUO; Int'l: RUO

TRUSTED_SUSTAINABILITY
Specifica

Specifica

Mucin 3 (MUC3) Ab-2
Monoclonal
200μg/mL
Mouse
100 μL
Cancer and Tumor Biology
Human
IgG2a
Immunohistochemistry (Paraffin)
1143/B7
Unconjugated
MUC3 tandem repeat peptide
RUO
Primary
Concentrated
Suggerimenti di prodotto

Suggerimenti di prodotto

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.