Learn More
Mucin 3 (MUC3) Ab-2 Mouse Monoclonal Antibody, Epredia™
Epredia™ Mucin 3 (MUC3) Ab-2, Mouse Monoclonal Antibody provides accurate, reproducible results.
Marca: Epredia MS-1734-P1ABX
Descrizione
Mucin 3 (MUC3) is a glycoprotein. Distributed in colon and rectum, and to a lesser extent in breast, lung and salivary gland tissues.Host Species: Mouse
Clone: 1143/B7
Isotype: IgG2a
Species Reactivity: Human. Others not known.
Epitope: Tandem repeats of the human MUC3
Immunogen: MUC3 tandem repeat peptide
Molecular Weight: High
Positive Control: Colon CA
Cellular Localization: Membrane
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
Note:
USA: RUO; Int'l: RUO
Specifica
| Mucin 3 (MUC3) Ab-2 | |
| Monoclonal | |
| 1mg/mL | |
| Mouse | |
| 100 μL | |
| Cancer and Tumor Biology | |
| Human | |
| IgG2a |
| Immunohistochemistry (Paraffin) | |
| 1143/B7 | |
| Unconjugated | |
| MUC3 tandem repeat peptide | |
| RUO | |
| Primary | |
| Concentrated |
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.