Learn More
Negative Control for Rabbit IgG Ab-1, Epredia™
Take advantage of a quality product that delivers accurate results with the Epredia™ Negative Control for Rabbit IgG Ab-1, Rabbit Polyclonal Antibody.
Marca: Epredia NC-100-P1ABX
Dettagli aggiuntivi : Peso : 1.00000kg
Descrizione
This IgG was purified from serum collected from a rabbit prior to immunization. It does not react with any antigen of human or animal cells. The serum was purified under the same experimental conditions as for the other rabbit polyclonal antibodies of NeoMarkers. In order to serve as a true negative control, this IgG should be used under the identical conditions and at the same concentration as NeoMarkers' other purified rabbit polyclonal IgG antibodies.Host Species: Rabbit
Isotype: IgG
Recommended for:
Negative control for:
- Immunohistochemistry (Frozen and formalin/paraffin)
- Western Blotting
- Immunofluorescence
- Immunoprecipitation
- Flow Cytometry
- ELISA
- Gel Supershift
Note:
USA: RUO; Int'l: RUO
Specifica
Negative Control for Rabbit IgG Ab-1 | |
Polyclonal | |
1mg/mL | |
Rabbit | |
RUO | |
Primary | |
Concentrated |
ELISA, Flow Cytometry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
Polyclonal | |
Unconjugated | |
500 μL | |
Negative Controls | |
All species | |
IgG |