missing translation for 'onlineSavingsMsg'
Learn More

Negative Control for Rabbit IgG Ab-1, Epredia™

Take advantage of a quality product that delivers accurate results with the Epredia™ Negative Control for Rabbit IgG Ab-1, Rabbit Polyclonal Antibody.

Marca:  Epredia NC-100-PABX

Dettagli aggiuntivi : Peso : 0.00180kg

 vedi altre versioni di questo prodotto

Codice prodotto. 12612667

Questo articolo non è attualmente disponibile o è stato dismesso.
La preghiamo di consultare la pagina del prodotto per individuare le possibili alternative.


Descrizione

Descrizione

This IgG was purified from serum collected from a rabbit prior to immunization. It does not react with any antigen of human or animal cells. The serum was purified under the same experimental conditions as for the other rabbit polyclonal antibodies of NeoMarkers. In order to serve as a true negative control, this IgG should be used under the identical conditions and at the same concentration as NeoMarkers' other purified rabbit polyclonal IgG antibodies.

Host Species: Rabbit

Isotype: IgG

Recommended for:

Negative control for:

  • Immunohistochemistry (Frozen and formalin/paraffin)
  • Western Blotting
  • Immunofluorescence
  • Immunoprecipitation
  • Flow Cytometry
  • ELISA
  • Gel Supershift

Note:

USA: RUO; Int'l: RUO

Specifica

Specifica

Negative Control for Rabbit IgG Ab-1
Polyclonal
1mg/mL
Rabbit
RUO
Primary
Concentrated
ELISA, Flow Cytometry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Polyclonal
Unconjugated
1 mL
Negative Controls
All species
IgG
Suggerimenti di prodotto

Suggerimenti di prodotto

Videos
SDS
Documenti

Documenti

Certificati
Offerte speciali

Offerte speciali