Learn More
Epredia™ Lab Vision™ p130/Rb2 Ab-2 Mouse Monoclonal Antibody with BSA and azide
Take advantage of a quality product that delivers accurate results with the Epredia™ p130/Rb2 Ab-2, Mouse Monoclonal Antibody.
Marca: Epredia™ MS-866-P1ABX
Dettagli aggiuntivi : Peso : 1.00000kg
Descrizione
Two retinoblastoma-related proteins, p107 and pRb2/p130 are structurally and functionally similar to the product of the retinoblastoma gene (pRb/p105). Like pRb they bind transforming proteins of DNA tumor viruses through a particular region called the “pocket domain”. Both proteins play a fundamental role in growth control. The highest percentage of undetectable levels and the tightest inverse correlation with the histological grading and with PCNA expression in the most aggressive tumor types were found for pRb2/p130, which may suggest an important role for this protein in the pathogenesis and progression of lung cancer.Host Species: Mouse
Clone: 130P215; same as DCS-215
Isotype: IgG1
Species Reactivity: Human and Rat. Others not known.
Epitope: aa 878-913
Immunogen: A synthetic peptide from p130 corresponding to aa 878-913
Molecular Weight: 130kDa
Positive Control: A427 cells, Tonsil
Cellular Localization: Nuclear
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
- Western Blotting
- Immunofluorescence
- Immunoprecipitation (Denatured verified)
Note:
USA: RUO; Int'l: RUO
Specifica
p130/Rb2 Ab-2 | |
Monoclonal | |
1mg/mL | |
Mouse | |
100 μL | |
Cell Cycle | |
Human, Rat | |
IgG1 |
Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
130P215; same as DCS-215 | |
Unconjugated | |
A synthetic peptide from p130 corresponding to aa878-913 | |
RUO | |
Primary | |
Concentrated |