Learn More
Epredia™ Lab Vision™ Secretory Component Ab-2 Mouse Monoclonal Antibody, 1.9mg/mL, BSA and azide
Provide accurate, reproducible results with the Epredia™ Secretory Component Ab-2, Mouse Monoclonal Antibody.
Marca: Epredia™ MS-1367-P0
Dettagli aggiuntivi : Peso : 1.00000kg
Descrizione
Secretory Component is an epithelial transport receptor for polymeric IgA and IgM. Antibody to secretory component has been shown to be useful for identifying glandular carcinomas.Host Species: Mouse
Clone: COMPO2; same as SC-05
Isotype: IgG1
Species Reactivity: Human and Rat. Others not known.
Epitope: Not determined
Immunogen: Semi-purified human secretory component (SC)
Molecular Weight: 80kDa
Positive Control: Small intestine, Colon CA, Stomach
Cellular Localization: Cytoplasmic
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
- Immunofluorescence
Note:
USA: RUO; Int'l: RUO
Specifica
Secretory Component Ab-2 | |
Monoclonal | |
1.9mg/mL | |
Mouse | |
100 μL | |
Cancer and Tumor Biology | |
Human, Rat | |
IgG1 |
Immunofluorescence, Immunohistochemistry (Paraffin) | |
COMPO2; same as SC-05 | |
Unconjugated | |
Semi- purified human secretory component (SC) | |
RUO | |
Primary | |
Concentrated |