missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ Lab Vision™ Secretory Component Ab-2 Mouse Monoclonal Antibody, 1.9mg/mL, BSA and azide

Provide accurate, reproducible results with the Epredia™ Secretory Component Ab-2, Mouse Monoclonal Antibody.

Marca:  Epredia™ MS-1367-P1

Dettagli aggiuntivi : Peso : 1.00000kg

 vedi altre versioni di questo prodotto

Codice prodotto. 12699656

  • 370.00€ / 500µL
Spedizione stimata: 20-05-2024
per visualizzare lo stock disponibile

Effettua il per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso!


Descrizione

Descrizione

Secretory Component is an epithelial transport receptor for polymeric IgA and IgM. Antibody to secretory component has been shown to be useful for identifying glandular carcinomas.

Host Species: Mouse

Clone: COMPO2; same as SC-05

Isotype: IgG1

Species Reactivity: Human and Rat. Others not known.

Epitope: Not determined

Immunogen: Semi-purified human secretory component (SC)

Molecular Weight: 80kDa

Positive Control: Small intestine, Colon CA, Stomach

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Immunofluorescence

Note:

USA: RUO; Int'l: RUO

Specifica

Specifica

Secretory Component Ab-2
Monoclonal
1.9mg/mL
Mouse
500 μL
Cancer and Tumor Biology
Human, Rat
IgG1
Immunofluorescence, Immunohistochemistry (Paraffin)
COMPO2; same as SC-05
Unconjugated
Semi- purified human secretory component (SC)
RUO
Primary
Concentrated
Suggerimenti di prodotto

Suggerimenti di prodotto

Videos
SDS
Documenti

Documenti

Certificati
Offerte speciali

Offerte speciali