Learn More
Epredia™ Lab Vision™ XPF Ab-1 Mouse Monoclonal Antibody, 200μg/mL, BSA and azide
Provide accurate, reproducible results with the Epredia™ XPF Ab-1, Mouse Monoclonal Antibody.
Marca: Epredia™ MS-1381-P1
Dettagli aggiuntivi : Peso : 0.00190kg
Descrizione
The structure-specific ERCC1/XPF endonuclease complex is implicated in the repair of two distinct types of lesions in DNA: NER for UV-induced lesions and bulky chemical adducts; and recombination repair of the very genotoxic interstrand cross-links. NER mechanism involves dual incisions on both sides of the damage catalyzed by two nucleases. In mammalian cells XPG cleaves 3' of the DNA lesion while the ERCC1-XPF complex makes the 5' incision.Host Species: Mouse
Clone: 219
Isotype: IgG2/κ
Species Reactivity: Human. Others not known.
Epitope: Not determined
Immunogen: Recombinant human XPF protein
Molecular Weight: ∽110kDa
Positive Control: MCF-7 cells or human tonsil
Cellular Localization: Nuclear
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
- Western Blotting
Note:
USA: RUO; Int'l: RUO
Specifica
XPF Ab-1 | |
Monoclonal | |
200μg/mL | |
Mouse | |
500 μL | |
DNA Repair | |
Human | |
IgG2 |
Immunohistochemistry (Paraffin), Western Blot | |
219 | |
Unconjugated | |
Recombinant human XPF protein | |
RUO | |
Primary | |
Concentrated |