Learn More
Labconco™ Automatic Pressure Controller Gas and Vacuum Tubing Connection Kit
For use with Precise™ Controlled Atmosphere Glove Boxes
Marca: Labconco™ 5245200
Dettagli aggiuntivi : Peso : 0.80000kg
Descrizione
Include the flexible vinyl tubing and fittings necessary to connect user-supplied inert gas supply tank and rotary vane vacuum pump to valves located on the glove box.
Specifica
20 lb. | |
9.1 Kg | |
Enclosure Kit | |
Precise™ Basic™ Glove Box |