Learn More
Labconco™ Fast-Freeze™ Flask Filter Paper
Fits into top of flask to prevent partial loss of product during freeze drying.
Marca: Labconco™ 7544810
Dettagli aggiuntivi : Peso : 0.80000kg
Descrizione
Fits into top of flask to prevent partial loss of product during freeze drying.
No complex filter retainer needed.
Specifica
Disc | |
prevents partial loss of product during freeze drying | |
1000 per pack | |
Flask Filter |