Learn More
Dostmann Elecronic Termometro a inserimento Labtherm
With a measuring range of 39.9°C to 249.9°C and an accuracy of ±0.5°C, this LabTherm digital catering thermometer is particularly suitable to measure liquids, food, etc.
Marca: Dostmann Elecronic 5020-0398
Dettagli aggiuntivi : Peso : 0.07200kg
Descrizione
- Labtherm Insertion thermometer
- Ergonomic shaped pocket thermometer with robust insertion tip
- The long sensor tube is particular suitable to measure in liquids, food etc
- LabTherm digital catering thermometer with a precise read-out of temperature over the range of -39.9°C to 249.9°C