Learn More
Brady™ Laminated Polyester Anti-skid Tape-Die-Cut Cleats
I nastri antisdrucciolo Anti-Skid contribuiscono a evitare scivolamenti e cadute su pavimenti bagnati o scivolosi. Il nastro Anti-Skid aderisce in modo permanente a pavimenti puliti e asciutti, resistendo a grasso e olio. Utilizzabile in interni ed esterni.
Marca: Brady™ 078195
Dettagli aggiuntivi : Peso : 2.50000kg
Descrizione
- Anti-Skid (anti-slip) tapes help to prevent slipping and falling on wet or slippery floors.
- The rugged polyester tape is adhesive backed and coated with thousands of particles of durable aluminium oxide grit.
- Anti-Skid tape bonds permanently to clean, dry floors, withstands grease and oil and will work indoors or outdoors.
- Anti-Skid tape is available in a variety of colors and sizes to suit all of your floor marking needs
Specifica
50 Pk. | |
Permanent Acrylic | |
Traction on steps, slick floors, and egress pathway marking | |
Polyester | |
600 x 150 mm | |
5.91 in. | |
Black | |
600 mm |
2 ft. | |
Anti-Skid Tape-Die-Cut Cleats - Black | |
Anti-skid Tape | |
-40°C to +70°C | |
0.162 mm | |
-40°F to +158°F | |
150 mm |