Learn More
Brady™ Laminated Polyester Anti-skid Tape-Die-Cut Cleats
I nastri antisdrucciolo Anti-Skid contribuiscono a evitare scivolamenti e cadute su pavimenti bagnati o scivolosi. Il nastro Anti-Skid aderisce in modo permanente a pavimenti puliti e asciutti, resistendo a grasso e olio. Utilizzabile in interni ed esterni.
Marca: Brady™ 078162
Dettagli aggiuntivi : Peso : 1.65560kg
Descrizione
- Anti-Skid (anti-slip) tapes help to prevent slipping and falling on wet or slippery floors.
- The rugged polyester tape is adhesive backed and coated with thousands of particles of durable aluminium oxide grit.
- Anti-Skid tape bonds permanently to clean, dry floors, withstands grease and oil and will work indoors or outdoors.
- Anti-Skid tape is available in a variety of colors and sizes to suit all of your floor marking needs
Specifica
24 Pk. | |
Permanent Acrylic | |
Traction on steps, slick floors, and egress pathway marking | |
Polyester | |
150 x 600 mm | |
24 in. | |
Photoluminescent, Black | |
150 mm |
5.9 in. | |
Glow-in-the-Dark Die-Cut Cleats - Black | |
Anti-skid Tape | |
-40°C to +70°C | |
0.162 mm | |
-40°F to +158°F | |
600 mm | |
ANSI A1264.2-2006, NSFI Certified for High Traction, OSHA 1910.144, OSHA 1910.22 |