missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Laminin Antibody (4C7), Novus Biologicals™
Mouse Monoclonal Antibody
Marca: Novus Biologicals NB120-17107-0.1ML
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
Laminin Monoclonal specifically detects Laminin in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifica
| Laminin | |
| Monoclonal | |
| Unconjugated | |
| Tissue culture supernatant, containing less than 0.1% sodium azide with Sodium Azide | |
| LAMA1 | |
| The A chain of human laminin. | |
| 0.1 mL | |
| Angiogenesis, Apoptosis, Cancer, Cell Biology, Cellular Markers, Extracellular Matrix, Neuroscience, Signal Transduction, Tumor Suppressors | |
| 284217 | |
| Human | |
| Supernatant |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| 4C7 | |
| Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500 | |
| LAMA, Laminin A chain, laminin subunit alpha-1, laminin, alpha 1, Laminin-1 subunit alpha, Laminin-3 subunit alpha, S-LAM alpha, S-laminin subunit alpha | |
| Mouse | |
| Tissue culture supernatant | |
| RUO | |
| Primary | |
| NB120-17107 reacts with the terminal globular domain of the A chain. The staining with this is sensitive to fixation of paraffin tissues and use of proteolytic enzymatic treatment. | |
| Store at 4°C. | |
| IgG2a κ |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
For Research Use Only
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto