Learn More
Brady™ LaserTab Self-laminating Polyester Labels
Blank, printable labels suitable for use with Laser Printers.
Marca: Brady™ 029740
Dettagli aggiuntivi : Peso : 0.40820kg
Descrizione
LaserTab Labels are available in the following formats:
- Tamper-evident polyolefin
- Polyester (UL Recognized Component when printed with designated laser printer toners)
- Self-laminating
- Nylon cloth
- Paper
- Polyimide
Specifica
Not Autoclavable | |
White, Transparent | |
Self-laminating Polyester | |
Non-sterile | |
Laser Printers | |
13 | |
5148 Labels | |
-70°C to +110°C | |
19.1 mm | |
12.7 mm | |
19.1 mm |
Permanent Acrylic | |
Matte | |
Laser Printing | |
Wire and Cable Labeling | |
15.2 mm | |
156 | |
Rectangle | |
0.051 mm | |
20 AWG to 12 AWG | |
12.7 mm | |
9.4 mm |