missing translation for 'onlineSavingsMsg'
Learn More

Piercan™ Latex Dry Box Gloves

Catalog Number p-8008469
Click to view available options
Diametro polso (sistema metrico):
156
186
220
300
Size:
7
9
Spessore (sistema imperiale):
15 mil
24 mil
This item is not returnable. View return policy
This item is not returnable. View return policy
Certificazione/Conformità CE 0086 Certified
Colore Natural
Finitura esterna Smooth
Materiale Latex
Product Type Dry Box Gloves
Size 7
Spessore (sistema imperiale) 15 mil
Da utilizzare con (applicazione) Ideal for use in Chemical & Medical Research, Pharmaceutical & Electronics Industries, Vapor Blasting
Stile polso guanto Beaded/Rolled
Dimensioni porta (sistema metrico) 200 mm
Quantità 2/Pk.
Resistente a To Most Acids, Alkalis and Organic Chemicals
Lunghezza (sistema metrico) 820 mm
Diametro polso (sistema metrico) 156
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'