Learn More
Emma Safety Footwear Leo XXD6T Safety Shoes
An orthopedic version of the standard Leo. Includes a steel toe cap, polyurethane outer toe cap and an extremely resilient reinforced sole. The ideal choice for those with slightly broader than normal feet or orthopedic requirements.
Marca: Emma Safety Footwear MM61002839
Dettagli aggiuntivi : Peso : 1.48500kg
Descrizione
- S3-Safety shoe with sole plate for wet work
- Full grain leather construction with Polyurethane outsole
- Hydro-Tec™ Sanitized Silver for drier, fresher feet
- Sanitized
Specifica
EN ISO 20345:2011 | |
Black | |
Hydro-Tec™ Sanitized Silver | |
39 |
Laces | |
Leo XXD6T | |
Leather | |
Steel |