missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LHX4 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 11183-1-AP-150UL
Additional Details : Peso : 0.00010kg
Description
This gene encodes a member of a large protein family which contains the LIM domain, a unique cysteine-rich zinc-binding domain. The encoded protein may function as a transcriptional regulator and be involved in control of differentiation and development of the pituitary gland. Mutations in this gene are associated with syndromic short stature and pituitary and hindbrain defects. An alternative splice variant has been described but its biological nature has not been determined.Specifications
LHX4 | |
Polyclonal | |
Unconjugated | |
LHX4 | |
Gsh 4, Gsh4, LHX4, LIM homeobox 4, LIM homeobox protein 4, LIM/homeobox protein Lhx4 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
16872, 360858, 89884 | |
-20°C | |
Liquid |
Western Blot | |
0.23 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P53776, Q969G2 | |
Lhx4 | |
LHX4 Fusion Protein Ag1642 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |