Learn More
Abeba™ Light 31056 Shoes
ESD dissipative shoe includes special respiration absorbing lining for the foot to breathe.
Marca: Abeba™ 31056/47
Dettagli aggiuntivi : Peso : 1.00000kg
Descrizione
- CE EN ISO 20345:2011 S1 SRA
- Leather with Air-Mesh textile inlays
- Black
- Steel toe cap
- Slip resistance PU outsole sole
- Replaceable acc Wave insole (article no. 3557)
- Sanitized treated
- ESD dissipative according to DIN EN 61340
- Breathable lining with Silver Point
- Instep area with lacing
- Reflective material
- Suitable for heel and foot lift
Specifica
CE EN ISO 20345:2011 S1 SRA | |
Leather | |
Slip-resistant | |
31056 |
Black | |
47 | |
Polyurethane | |
Steel |