Learn More
Abeba™ Light 31058 Shoes
ESD dissipative shoe includes special respiration absorbing lining for the foot to breathe.
Marque:  Abeba™ 31058/41
Informations supplémentaires : Peso : 1.11900kg
 Afficher plus de versions de ce produit
Code produit. 15648467
Questo articolo non è attualmente disponibile o è stato dismesso.
La preghiamo di consultare la pagina del prodotto per individuare le possibili alternative.
Description
Description
  • CE EN ISO 20345:2011 S1 SRA
  • Leather with Air-Mesh textile inlays
  • White
  • Steel toe cap
  • Slip resistance PU outsole sole
  • Replaceable acc Wave insole (article no. 3557)
  • Sanitized treated
  • ESD dissipative according to DIN EN 61340
  • Breathable lining with Silver ions
  • Instep area with lacing
Spécification
Spécification
CE EN ISO 20345:2011 S1 SRA
Leather
Slip-resistant
31058
White
41
Polyurethane
Steel
Suggestions de produits
Suggestions de produits
Vidéos
FDS
Documents
Documents
Certificats
Promotions
Promotions