Learn More
Abeba™ Light 31058 Shoes
ESD dissipative shoe includes special respiration absorbing lining for the foot to breathe.
Marca: Abeba™ 31058/46
Dettagli aggiuntivi : Peso : 1.11900kg
Descrizione
- CE EN ISO 20345:2011 S1 SRA
- Leather with Air-Mesh textile inlays
- White
- Steel toe cap
- Slip resistance PU outsole sole
- Replaceable acc Wave insole (article no. 3557)
- Sanitized treated
- ESD dissipative according to DIN EN 61340
- Breathable lining with Silver ions
- Instep area with lacing
Specifica
CE EN ISO 20345:2011 S1 SRA | |
Leather | |
Slip-resistant | |
31058 |
White | |
46 | |
Polyurethane | |
Steel |