Learn More
Abeba™ Light 31120 Shoes
ESD dissipative shoe includes special respiration absorbing lining for the foot to breathe.
Marca: Abeba™ 31120/40
Dettagli aggiuntivi : Peso : 1.04000kg
Descrizione
- CE EN ISO 20347:2012 O1 FO SRA
- Smooth leather, perforated
- White
- Slip resistance PU outsole sole
- Replaceable acc Wave insole (article no. 3559)
- Sanitized treated
- ESD dissipative according to DIN EN 61340
- Breathable lining with Silver ions
- Double velcro fastening
- Reflective material
- Suitable for heel and foot lift
Specifica
CE EN ISO 20347:2012 O1 FO SRA | |
Leather | |
Slip-resistant | |
31120 |
White | |
40 | |
Polyurethane |