Learn More
Abeba™ Light 31129 Shoes
ESD dissipative shoe includes special respiration absorbing lining for the foot to breathe.
Marca: Abeba™ 31129/44
Dettagli aggiuntivi : Peso : 0.90000kg
Descrizione
- CE EN ISO 20347:2012 O2 FO SRA
- Microfiber black
- Slip resistance PU outsole sole
- Replaceable acc Wave insole (article no. 3559)
- Sanitized treated
- ESD dissipative according to DIN EN 61340
- Breathable lining with Silver ions
- Instep area with elastic band
Specifica
CE EN ISO 20347:2012 O2 FO SRA | |
Microfiber | |
Slip-resistant | |
31129 |
Black | |
44 | |
Polyurethane |