Learn More
Abeba™ Light 31131 Shoes
ESD dissipative shoe includes special respiration absorbing lining for the foot to breathe.
Marca: Abeba™ 31131/47
Dettagli aggiuntivi : Peso : 1.00000kg
Descrizione
- CE EN ISO 20347:2012 O1 FO SRA
- Leather with Air-Mesh textile inlays
- White
- Slip resistance PU outsole sole
- Replaceable acc Wave insole (article no. 3559)
- Sanitized treated
- ESD dissipative according to DIN EN 61340
- Breathable lining with Silver ions
- Double velcro fastening
Specifica
CE EN ISO 20347:2012 O1 FO SRA | |
Leather | |
Slip-resistant | |
31131 |
White | |
47 | |
Polyurethane |