Learn More
Abeba™ Light 31163 Shoes
ESD dissipative shoe includes special respiration absorbing lining for the foot to breathe.
Marca: Abeba™ 31163/40
Dettagli aggiuntivi : Peso : 0.60000kg
Descrizione
- CE EN ISO 20347:2012 OB A, E, FO SRA
- Smooth leather white/light blue
- Slip resistance PU outsole sole
- Replaceable acc Wave insole (article no. 3558)
- Sanitized treated
- ESD dissipative according to DIN EN 61340
- Heel strap non-foldable, adjustable
- Breathable lining with Silver ions
- Instep area with lacing
- velcro fastening
- Reflective material
Specifica
CE EN ISO 20347:2012 OB A, E, FO SRA | |
Leather | |
Slip-resistant | |
31163 |
White | |
40 | |
Polyurethane |