Learn More
Abeba™ Light 31853 Shoes
ESD dissipative shoe includes special respiration absorbing lining for the foot to breathe.
Marca: Abeba™ 31853/35
Dettagli aggiuntivi : Peso : 1.32700kg
Descrizione
- CE EN ISO 20345:2011 S3 SRA
- Smooth leather
- Black in ATEX design
- Steel toe cap
- Metalfree penetration resistance
- Slip resistance PU outsole sole
- Replaceable acc Wave insole (article no. 3557)
- Sanitized treated
- ESD dissipative according to DIN EN 61340
- Breathable lining with Silver Point
- Instep area with lacing
- Reflective material
- Suitable for heel and foot lift
Specifica
CE EN ISO 20345:2011 S3 SRA | |
Leather | |
Slip-resistant | |
31853 |
Black | |
35 | |
Polyurethane | |
Steel |