missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Ligustrazine hydrochloride
Click to view available options
Quantità:
1 g
5 g
Descrizione
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifica
Materiale o nome chimico | Ligustrazine hydrochloride |
Quantità | 1 g |
Punti di ebollizione | 193.0°C (730.0 mmHg) |
Peso formulazione | 172.66 |
InChI Key | RQKFOGXUTRDQPB-UHFFFAOYSA-N |
SMILES | Cl.CC1=NC(C)=C(C)N=C1C |
IUPAC Name | tetramethylpyrazine hydrochloride |
Molecular Weight (g/mol) | 172.66 |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'