Learn More
BD Vacutainer Lithium Heparin Tubes
Plasma tube for clinical chemistry includes spray-dried lithium heparin as an anticoagulant.
Marca: BD Vacutainer 368886
Dettagli aggiuntivi : Peso : 1.00000kg
Descrizione
- Plasma tubes for clinical chemistry with spray-dried lithium heparin.
- Heparin acts as an anticoagulant as it develops an antithrombin complex.
- Plasma tubes should be gently inverted 180° and back 8-10 times.
- Correct mixing immediately after the blood sample has been taken is extremely important to avoid microclotting.
- The lithium heparin is spray dried onto the inner walls of the tubes using a special procedure so that the additive is evenly distributed to achieve the best possible solubility.
- For clinical chemistry, lithium heparin is generally preferred over sodium heparin.
Specifica
Lithium Heparin | |
6 mL | |
13 x 100 mm |
PET | |
Green | |
Paper |