Learn More
Lithium ICP & ICP-MS Standard, 100 mg/L Li in HNO3 2%, Honeywell Fluka™
Marca: Honeywell Fluka E1329-100ML
Dettagli aggiuntivi : Peso : 0.10000kg Transport : UN number : 3264 Chem class : 8 Pack group : III
Descrizione
Honeywell Fluka™ analytical standards offer a growing range of high quality inorganic and organic reference materials for the European market produced according to ISO 9001. The products are suitable for a wide range of industries and applications and come complete with a detailed Certificate of Analysis (CoA) in every delivery, and are eligible for free expert technical support ensuring you have the information you need to quickly start your analysis and pass an audit.
Specifica
Lithium ICP and ICP-MS Standard | |
Standards for ICP and ICP-MS | |
<2 | |
100 mL |