Learn More
Lithium tetrafluoroborate, 98%, pure, anhydrous, Thermo Scientific Chemicals
54.70€ - 181.00€
Identificatori chimici
CAS | 14283-07-9 |
---|---|
Formula molecolare | BF4Li |
Molecular Weight (g/mol) | 93.74 |
Numero MDL | MFCD00011087 |
InChI Key | UFXJWFBILHTTET-UHFFFAOYSA-N |
Sinonimo | lithium tetrafluoroborate, lithium fluoroborate, lithium tetra fluoroborate, lithium 1+ ion tetrafluoroborate, lithium borofluoride, unii-af751cnk2n, libf4, lithotab tetrafluoroborate, acmc-209cp3, af751cnk2n |
PubChem CID | 4298216 |
IUPAC Name | lithium(1+) tetrafluoroboranuide |
SMILES | [Li+].F[B-](F)(F)F |
Codice del prodotto | Marca | Quantità | Imballaggio | Prezzo | Quantità e disponibilità | ||||
---|---|---|---|---|---|---|---|---|---|
Codice del prodotto | Marca | Quantità | Imballaggio | Prezzo | Quantità e disponibilità | ||||
10032880
|
Thermo Scientific Acros
223770100 |
10 g | Plastic bottle |
54.70€
10g |
Effettua il login per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso! | ||||
10358550
|
Thermo Scientific Acros
223770500 |
50 g | Plastic bottle |
181.00€
50g |
Effettua il login per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso! | ||||
Descrizione
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Identificatori chimici
14283-07-9 | |
93.74 | |
UFXJWFBILHTTET-UHFFFAOYSA-N | |
4298216 | |
[Li+].F[B-](F)(F)F |
BF4Li | |
MFCD00011087 | |
lithium tetrafluoroborate, lithium fluoroborate, lithium tetra fluoroborate, lithium 1+ ion tetrafluoroborate, lithium borofluoride, unii-af751cnk2n, libf4, lithotab tetrafluoroborate, acmc-209cp3, af751cnk2n | |
lithium(1+) tetrafluoroboranuide |
Specifica
Lithium tetrafluoroborate | |
98% | |
Gray to White | |
98% | |
LiBF4 | |
05,416; 10,248 | |
lithium tetrafluoroborate, lithium fluoroborate, lithium tetra fluoroborate, lithium 1+ ion tetrafluoroborate, lithium borofluoride, unii-af751cnk2n, libf4, lithotab tetrafluoroborate, acmc-209cp3, af751cnk2n | |
UFXJWFBILHTTET-UHFFFAOYSA-N | |
lithium(1+) tetrafluoroboranuide | |
4298216 | |
98% | |
0.852 |
275°C | |
14283-07-9 | |
Crystalline Powder and/or Crystals | |
BF4Li | |
MFCD00011087 | |
15, 5599 | |
Solubility in water: soluble | |
[Li+].F[B-](F)(F)F | |
93.74 | |
93.75 | |
Pure | |
0.8520g/mL |
Sicurezza e movimentazione
GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if inhaled.
Harmful if swallowed.
GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 238-178-9
missing translation for 'tsca' : TSCA
- Lithium tetrafluoroborate, anhydrous
Avvertenza
- Pericolo
Categoria di pericolo
- Tossicità acuta Categoria 4
- Lesione oculare grave/irritazione oculare Categoria 2
- Mutagenicità di cellule germinali Categoria 2
- Corrosione/irritazione della pelle Categoria 2
Indicazioni di pericolo
- H315-Provoca irritazione cutanea.
- H319-Provoca grave irritazione oculare.
- H341-Sospettato di provocare alterazioni genetiche.
- H302-Nocivo se ingerito.
Consigli di prudenza
- P280-Indossare guanti/indumenti protettivi/Proteggere gli occhi/il viso.
- P301+P312-IN CASO DI INGESTIONE accompagnata da malessere: contattare un CENTRO ANTIVELENI/medico/
- P301+P330+P331-IN CASO DI INGESTIONE: sciacquare la bocca. NON provocare il vomito.
- P304+P340-IN CASO DI INALAZIONE: trasportare l'infortunato all'aria aperta e mantenerlo a riposo in posizione che favorisca la respirazione.
- P305+P351+P338-IN CASO DI CONTATTO CON GLI OCCHI: sciacquare accuratamente per parecchi minuti. Togliere le eventuali lenti a contatto se è agevole farlo. Continuare a sciacquare.
- P310-Contattare immediatamente un CENTRO ANTIVELENI/un medico .
RUO – Research Use Only