Learn More
Notrax Logo Washable™ Custom Printed Logo Mat
Logo Washable™ can be fully customized with BrilliantStep™ state-of-the-art printing technology allowing 3-dimensional effects, fine lines and color shading, for each mat to have its truly individual image. Nitrile rubber backing is washable up to 50°C.
Marca: Notrax 190S0035WH
Dettagli aggiuntivi : Peso : 3.60000kg
Descrizione
- Functional and washable logo entrance mat that can be completely personalized using BrilliantStep™ full color printing with almost infinite color combinations
- The open Frieze tuft gives true depth of color and a trackless surface making it more durable as well as more effective in hiding dirt and foot traffic
- Nitrile rubber backing is washable up to 50°C with cleated pattern to minimize movement on all surfaces and to keep the mat elevated on hard floors to allow drying underneath
- Logo Washable™ can be fully customized with BrilliantStep™ state-of-the-art printing technology allowing 3-dimensional effects, fine lines and color shading, for each mat to have its truly individual image
Specifica
59 in. | |
Industrial and Commercial Matting | |
Polyamide 6.6 High Twist Yarns | |
150 cm |
White | |
Frieze | |
85 cm |