Learn More
Lovibond™ Gardner Color Scale Based on Comparator System 2000
A flexible, modular system for visual color grading.
Marca: Lovibond™ 433340
Dettagli aggiuntivi : Peso : 2.12000kg
Descrizione
- Type: AF 334
- Range: 1-18 units
- Includes: Lovibond Comparator 2000+ with Daylight 2000 Lighting Unit, Gardner discs 4/30 AS and 4/30 BS, and (2) W680/OG/10mm path length fused glass cells
Compliance
Conforms to ASTM D1544, ISO4630 Standards
Specifica
Conforms to ASTM D1544, ISO4630 Standards | |
Gardner Color Scale | |
Lovibond Comparator 2000+ with Daylight 2000 Lighting Unit, Gardner discs 4/30 AS and 4/30 BS, and (2) W680/OG/10mm path length fused glass cells | |
1 to 18 units |
Gardner Color Scale Based on Comparator System 2000 | |
A flexible, modular system for visual color grading. | |
AF 334 |