Learn More
Porcelaines Avignon™ Crogioli bassi in porcellana
Heat and cool substances in the laboratory using Porcelaines Avignon™ Low Form Porcelain Crucibles. They are available in various capacities.
Marca: Porcelaines Avignon™ 11308
Dettagli aggiuntivi : Peso : 0.45000kg
Descrizione
- Low walls
- Withstand high temperatures and rapid temperature changes
- Porcelain
Specifica
Porcellana | |
80 mm | |
Crogiolo |
130 ml | |
50 mm |