Learn More
LRRC18 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 24571-1-AP-150UL
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
May be involved in the regulation of spermatogenesis and sperm maturation.Specifica
| LRRC18 | |
| Polyclonal | |
| Unconjugated | |
| LRRC18 | |
| LRRC18, UNQ933, UNQ9338, VKGE9338 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 474354, 67580 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 0.39 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q8N456, Q9CQ07 | |
| LRRC18 | |
| LRRC18 Fusion Protein Ag20996 | |
| 150 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |