missing translation for 'onlineSavingsMsg'
Learn More

LSM7 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marca:  Proteintech 18941-1-AP-20UL

Dettagli aggiuntivi : Peso : 0.00010kg

 vedi altre versioni di questo prodotto

Codice prodotto. 16898754

  • 140.00€ / 20µL
Spedizione stimata: 15-04-2024
per visualizzare lo stock disponibile

Effettua il per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso!


Descrizione

Descrizione

Sm-like proteins were identified in a variety of organisms based on sequence homology with the Sm protein family. Sm-like proteins contain the Sm sequence motif, which consists of 2 regions separated by a linker of variable length that folds as a loop. The Sm-like proteins are thought to form a stable heteromer present in tri-snRNP particles, which are important for pre-mRNA splicing.
Specifica

Specifica

LSM7
Polyclonal
Unconjugated
LSM7
LSM7, YNL147W
Rabbit
Antigen Affinity Chromatography
RUO
362829, 51690, 66094
-20°C
Liquid
Western Blot, Immunohistochemistry (Paraffin)
0.2 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q9CQQ8, Q9UK45
LSM7
LSM7 Fusion Protein Ag13498
20 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Suggerimenti di prodotto

Suggerimenti di prodotto

Videos
SDS
Documenti

Documenti

Certificati
Offerte speciali

Offerte speciali