missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LY6D Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 17361-1-AP-150UL
Dettagli aggiuntivi : Peso : 0.00010kg
Descrizione
Granulocytes are a category of white blood cells characterized by the presence of granules in their cytoplasm. They are also called polymorphonuclear leukocytes (PMN or PML) because of the varying shapes of the nucleus, which is usually lobed into three segments. In common parlance, the term polymorphonuclear leukocyte often refers specifically to neutrophil granulocytes, the most abundant of the granulocytes. Granulocytes or PMN are released from the bone marrow by the regulatory complement proteins.Specifica
LY6D | |
Polyclonal | |
Unconjugated | |
LY6D | |
E48, E48 antigen, Ly 6D, LY6D, Lymphocyte antigen 6D | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
17068, 8581 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.28 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P35459, Q14210 | |
Ly6d | |
LY6D Fusion Protein Ag10714 | |
150 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |