Learn More
3M™ Macadamia Protein ELISA Kit, E96MAC
Intended for the detection of macadamia proteins in clean-in-place water (CIP) final rinse water, environmental swab samples, food ingredients and processed food products.
Marca: 3M™ 7100151368
Dettagli aggiuntivi : Peso : 0.03200kg
Descrizione
- Designed to detect both processed and unprocessed target allergen proteins.
- Test can accommodate environmental swab, CIP rinse water and food samples.
- A universal extraction protocol streamlines testing for multiple analytes, saving you reagents and valuable time.
Specifica
Test | |
50 Minutes |