missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva Life Sciences™ Magnetic Filter Funnels
Unique magnetic seal allows easy, one-handed vacuum filtration of liquids.
Marca: Cytiva Life Sciences™ 4242-N
Dettagli aggiuntivi : Peso : 0.01100kg
Descrizione
- Magnetic filter funnel features a magnetic closure action with magnets that are enclosed in a durable base and funnel to provide an integral, leak-proof seal.
- Simple one-handed operation for vacuum filtration makes this essential tool a favorite of microbiologists by promoting asceptic laboratory handling techniques.
- The funnel features a sturdy construction that is shatterproof and can withstand multiple autoclave cycles.
- Polyphenylsulfone construction is compatible with anti-foaming agents and many other solvents.
- Convenient. 150 mL size allows for easy fit into small autoclaves and 500 mL size is ideal for filtration of large samples.
- Sturdy and safe.
- Polyphenylsulfone construction provides durability and added safety at a cost less than most glass funnels.
- Forceps access point allows easy filter retrieval.
- Graduated at 50 mL increments for accurate sample measurement.
- New 47 mm Sentino™ Magnetic Filter Funnels also available.
- #8 stopper with proper bore hole size included.
Specifica
300 mL | |
1 Ea. | |
Magnetic Filtering Funnel |