Learn More
Fisherbrand™ Manual Crimper, Steel, for Flip Top/Flip Off Seals or Flip Tear Up Seals
Crimping tools provide a reproducible, secure vial closure.
Marca: Fisherbrand™
Dettagli aggiuntivi : Peso : 0.60000kg
Descrizione
- Crimper
- Manual
- Steel
- Non-autoclavable
- Adjustable
Specifica
13mm Crimper | |
Steel | |
For Flip Tear Up |
13 mm | |
Not Autoclavable |