missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MARCH5 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 12213-1-AP-20UL
Dettagli aggiuntivi : Peso : 0.00010kg
Descrizione
MARCH5 is a ubiquitin ligase of the mitochondrial outer membrane that plays a role in the control of mitochondrial morphology by regulating mitofusin-2.Specifica
MARCH5 | |
Polyclonal | |
Unconjugated | |
MARCH5 | |
MARCH V, MARCH5, Mitochondrial ubiquitin ligase, MITOL, RING finger protein 153, RNF153 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
54708 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Immunoprecipitation | |
0.18 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q9NX47 | |
MARCH5 | |
MARCH5 Fusion Protein Ag2806 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |